Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osi17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275377.1 | NP_001262306.1 | 3 - 4 | 2192 | 104 | |
NM_001275378.1 | NP_001262307.1 | 2 - 3 | 1479 | 104 | ||
NM_001275379.1 | NP_001262308.1 | 3 - 4 | 2192 | 104 | ||
NM_141381.3 | NP_649638.2 | 2 - 3 | 1479 | 104 | ||
GenBank mRNA | AY118624.1 | - | 2 - 3 | 1476 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |