Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sunz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141416.6 | NP_649673.1 | 2 - 3 | 553 | 93 | |
GenBank mRNA | AY075298.1 | - | 2 - 3 | 534 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |