Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD98hc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275425.1 | NP_001262354.2 | 5 - 6 | 464 | 68 | |
NM_001275426.1 | NP_001262355.1 | 5 - 6 | 537 | 68 | ||
NM_001275427.1 | NP_001262356.1 | 2 - 3 | 387 | 68 | ||
NM_141495.3 | NP_649752.2 | 2 - 3 | 387 | 68 | ||
GenBank mRNA | BT044083.1 | - | 2 - 3 | 384 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |