Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141744.3 | NP_650001.2 | 1 - 2 | 179 | 57 | |
GenBank mRNA | BT050573.1 | - | 1 - 2 | 164 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |