Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316502.1 | NP_001303431.1 | 1 | 989 | 138 | |
NM_141791.3 | NP_650048.1 | 1 | 989 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |