Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260146.2 | NP_001247075.1 | 8 - 9 | 5848 | 79 | |
NM_141936.2 | NP_650193.1 | 8 - 9 | 5842 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |