Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141983.3 | NP_650240.1 | 3 - 4 | 1386 | 73 | |
NM_169482.1 | NP_731749.3 | 4 - 5 | 1663 | 73 | ||
GenBank mRNA | AF218811.1 | - | 3 - 4 | 1355 | 73 | |
BT100045.1 | - | 3 - 4 | 1318 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |