Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat89Ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142273.4 | NP_650530.2 | 5 - 6 | 3260 | 68 | |
GenBank mRNA | BT001511.1 | - | 5 - 6 | 3234 | 68 | |
BT016113.1 | - | 5 - 6 | 3232 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |