Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144602.2 | NP_001138074.1 | 2 - 3 | 644 | 61 | |
NM_001275775.1 | NP_001262704.1 | 2 - 3 | 741 | 61 | ||
NM_001275776.1 | NP_001262705.1 | 2 - 3 | 667 | 61 | ||
NM_001275777.1 | NP_001262706.1 | 2 - 3 | 667 | 61 | ||
NM_001275778.1 | NP_001262707.1 | 2 - 3 | 777 | 61 | ||
NM_142453.4 | NP_650710.3 | 2 - 3 | 777 | 61 | ||
NM_169803.4 | NP_732325.3 | 2 - 3 | 667 | 61 | ||
NM_169804.4 | NP_732326.3 | 2 - 3 | 676 | 61 | ||
NM_169805.4 | NP_732327.3 | 2 - 3 | 695 | 61 | ||
GenBank mRNA | AY118312.1 | - | 2 - 3 | 777 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |