Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104363.2 | NP_001097833.1 | 2 - 3 | 1084 | 77 | |
NM_001275789.1 | NP_001262718.1 | 2 - 3 | 2250 | 77 | ||
NM_001275790.1 | NP_001262719.1 | 4 - 5 | 2081 | 77 | ||
NM_142504.4 | NP_650761.2 | 4 - 5 | 2673 | 77 | ||
NM_169839.3 | NP_732384.1 | 4 - 5 | 2081 | 77 | ||
NM_169840.3 | NP_732385.1 | 3 - 4 | 1174 | 77 | ||
NM_169841.2 | NP_732386.1 | 2 - 3 | 794 | 77 | ||
GenBank mRNA | BT058039.1 | - | 2 - 3 | 794 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |