Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142639.4 | NP_650896.2 | 3 - 4 | 253 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |