Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142667.3 | NP_650924.3 | 8 - 9 | 1323 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |