Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142685.3 | NP_650942.1 | 1 - 2 | 154 | 70 | |
GenBank mRNA | BT003492.1 | - | 1 - 2 | 146 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |