Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CheB93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142713.3 | NP_650970.2 | 2 - 3 | 442 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |