Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142869.5 | NP_651126.3 | 2 - 3 | 527 | 69 | |
GenBank mRNA | AY051923.1 | - | 2 - 3 | 492 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |