Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golgin84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142993.3 | NP_651250.2 | 1 - 2 | 1395 | 64 | |
GenBank mRNA | AY071645.1 | - | 1 - 2 | 1380 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |