Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cad96Ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143092.3 | NP_651349.1 | 8 - 9 | 2022 | 60 | |
GenBank mRNA | AY070958.1 | - | 8 - 9 | 2020 | 60 | |
BT001657.1 | - | 8 - 9 | 2020 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |