Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276040.1 | NP_001262969.1 | 3 - 4 | 891 | 75 | |
NM_143161.2 | NP_651418.1 | 3 - 4 | 939 | 75 | ||
NM_170237.2 | NP_733116.1 | 3 - 4 | 1039 | 75 | ||
NM_170238.2 | NP_733117.1 | 3 - 4 | 975 | 75 | ||
GenBank mRNA | AY113254.1 | - | 3 - 4 | 1039 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |