Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GluRIID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143725.3 | NP_651982.1 | 6 - 7 | 1445 | 77 | |
GenBank mRNA | AJ251888.1 | - | 6 - 7 | 1402 | 77 | |
AJ251889.1 | - | 6 - 7 | 1402 | 77 | ||
BT150368.1 | - | 6 - 7 | 1445 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |