Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014616.2 | NP_001014616.2 | 3 | 1137 | 58 | |
GenBank mRNA | BT057996.1 | - | 3 | 1137 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |