Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169080.3 | NP_730966.1 | 4 - 5 | 1648 | 63 | |
GenBank mRNA | BT025208.2 | - | 4 - 5 | 1620 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |