Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: put | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275645.1 | NP_001262574.1 | 2 - 3 | 688 | 82 | |
NM_001275646.1 | NP_001262575.1 | 2 - 3 | 688 | 82 | ||
NM_169591.3 | NP_731926.1 | 2 - 3 | 688 | 82 | ||
GenBank mRNA | L38495.1 | - | 2 - 3 | 661 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |