Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104344.2 | NP_001097814.1 | 5 - 6 | 692 | 66 | |
NM_001144593.2 | NP_001138065.1 | 4 - 5 | 600 | 66 | ||
NM_001144594.2 | NP_001138066.1 | 4 - 5 | 653 | 66 | ||
NM_001275716.1 | NP_001262645.1 | 4 - 5 | 653 | 66 | ||
NM_169724.2 | NP_732164.1 | 4 - 5 | 653 | 66 | ||
NM_169725.2 | NP_732165.1 | 4 - 5 | 600 | 66 | ||
NM_169726.2 | NP_732166.1 | 4 - 5 | 600 | 66 | ||
GenBank mRNA | BT044209.1 | - | 4 - 5 | 631 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |