Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275759.1 | NP_001262688.1 | 9 - 10 | 2757 | 56 | |
NM_080105.4 | NP_524844.6 | 9 - 10 | 2830 | 56 | ||
NM_169781.4 | NP_732282.6 | 9 - 10 | 3944 | 56 | ||
NM_169782.3 | NP_732283.5 | 9 - 10 | 2885 | 56 | ||
NM_169783.4 | NP_732284.5 | 9 - 10 | 2830 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |