Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oamb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275845.2 | NP_001262774.1 | 5 - 6 | 1481 | 65 | |
NM_001275846.2 | NP_001262775.1 | 5 - 6 | 1481 | 65 | ||
NM_001316500.1 | NP_001303429.1 | 5 - 6 | 1481 | 65 | ||
NM_079930.4 | NP_524669.2 | 5 - 6 | 1481 | 65 | ||
NM_169913.2 | NP_732541.1 | 5 - 6 | 1481 | 65 | ||
NM_169914.2 | NP_732542.1 | 4 - 5 | 1055 | 65 | ||
GenBank mRNA | AJ007617.1 | - | 3 - 4 | 944 | 65 | |
BT010103.1 | - | 5 - 6 | 1445 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |