Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104378.2 | NP_001097848.2 | 5 - 6 | 1409 | 119 | |
NM_001104379.2 | NP_001097849.1 | 5 - 6 | 1332 | 119 | ||
NM_001275847.1 | NP_001262776.1 | 6 - 7 | 1419 | 119 | ||
NM_001275849.1 | NP_001262778.1 | 6 - 7 | 1419 | 119 | ||
NM_001275851.1 | NP_001262780.1 | 5 - 6 | 2744 | 119 | ||
NM_001275852.1 | NP_001262781.1 | 5 - 6 | 1158 | 119 | ||
NM_001275853.1 | NP_001262782.1 | 5 - 6 | 1409 | 119 | ||
NM_001275854.1 | NP_001262783.1 | 5 - 6 | 1158 | 119 | ||
NM_001275855.1 | NP_001262784.1 | 5 - 6 | 2744 | 119 | ||
NM_001369986.1 | NP_001356920.1 | 6 - 7 | 1419 | 119 | ||
NM_001369987.1 | NP_001356921.1 | 6 - 7 | 1419 | 119 | ||
NM_001369988.1 | NP_001356922.1 | 5 - 6 | 1158 | 119 | ||
NM_142656.4 | NP_650913.2 | 5 - 6 | 2744 | 119 | ||
NM_169925.3 | NP_732555.1 | 5 - 6 | 1158 | 119 | ||
NM_169926.2 | NP_732556.1 | 5 - 6 | 1158 | 119 | ||
NM_169927.3 | NP_732557.2 | 6 - 7 | 1419 | 119 | ||
NM_169929.3 | NP_732559.1 | 5 - 6 | 1409 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |