Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043281.2 | NP_001036746.1 | 7 - 8 | 4397 | 74 | |
NM_170037.2 | NP_732790.3 | 7 - 8 | 4430 | 74 | ||
GenBank mRNA | AB190214.1 | - | 7 - 8 | 3803 | 74 | |
BT099586.1 | - | 7 - 8 | 4384 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |