Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KrT95D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079749.6 | NP_524473.2 | 5 - 6 | 2573 | 73 | |
NM_170108.4 | NP_732933.1 | 6 - 7 | 1893 | 73 | ||
NM_170109.2 | NP_732934.2 | 3 - 4 | 909 | 73 | ||
GenBank mRNA | AY061587.1 | - | 4 - 5 | 797 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |