Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170175.4 | NP_733051.2 | 2 - 3 | 498 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |