Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc98D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170288.1 | NP_001163759.1 | 37 - 38 | 14286 | 71 | |
NM_001276107.1 | NP_001263036.1 | 31 - 32 | 11968 | 71 | ||
NM_079817.3 | NP_524541.3 | 38 - 39 | 14307 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |