Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkc98E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170296.2 | NP_001163767.1 | 7 - 8 | 1047 | 64 | |
NM_001300648.1 | NP_001287577.1 | 8 - 9 | 1113 | 64 | ||
NM_079821.4 | NP_524545.2 | 8 - 9 | 1079 | 64 | ||
GenBank mRNA | BT023513.1 | - | 7 - 8 | 1016 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |