Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079859.4 | NP_524598.1 | 3 - 4 | 598 | 100 | |
GenBank mRNA | BT010108.2 | - | 3 - 4 | 675 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |