Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079859.4 | NP_524598.1 | 5 - 6 | 827 | 70 | |
GenBank mRNA | BT010108.2 | - | 5 - 6 | 904 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |