Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276205.1 | NP_001263134.1 | 13 - 14 | 4105 | 57 | |
NM_001276206.1 | NP_001263135.1 | 13 - 14 | 4117 | 57 | ||
NM_001276207.1 | NP_001263136.1 | 13 - 14 | 4096 | 57 | ||
NM_001276208.1 | NP_001263137.1 | 13 - 14 | 4117 | 57 | ||
NM_079866.3 | NP_524605.1 | 13 - 14 | 4126 | 57 | ||
GenBank mRNA | BT126010.1 | - | 13 - 14 | 3904 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |