Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FeCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079874.3 | NP_524613.1 | 3 - 4 | 605 | 86 | |
NM_170579.3 | NP_733458.1 | 3 - 4 | 605 | 86 | ||
GenBank mRNA | AY058251.1 | - | 3 - 4 | 547 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |