Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275743.2 | NP_001262672.1 | 1 - 2 | 382 | 72 | |
NM_001275744.1 | NP_001262673.1 | 1 - 2 | 382 | 72 | ||
NM_079915.3 | NP_524654.2 | 1 - 2 | 382 | 72 | ||
GenBank mRNA | BT031265.1 | - | 1 - 2 | 293 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |