Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276024.1 | NP_001262953.1 | 4 - 5 | 1518 | 63 | |
NM_001276025.1 | NP_001262954.1 | 4 - 5 | 1518 | 63 | ||
NM_080146.3 | NP_524885.1 | 4 - 5 | 1518 | 63 | ||
NM_170654.2 | NP_733102.1 | 4 - 5 | 1518 | 63 | ||
NM_170655.2 | NP_733103.1 | 4 - 5 | 1518 | 63 | ||
NM_170656.2 | NP_733104.1 | 3 - 4 | 1453 | 63 | ||
NM_206570.2 | NP_996293.1 | 5 - 6 | 1574 | 63 | ||
NM_206571.2 | NP_996294.2 | 4 - 5 | 1518 | 63 | ||
GenBank mRNA | AY069590.1 | - | 4 - 5 | 1500 | 63 | |
L12372.1 | - | 4 - 5 | 1427 | 63 | ||
L12375.1 | - | 4 - 5 | 1493 | 63 | ||
L12376.1 | - | 3 - 4 | 1407 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |