Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GstD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080176.3 | NP_524915.1 | 1 | 617 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |