Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spdo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170493.3 | NP_733372.1 | 1 - 2 | 1498 | 81 | |
GenBank mRNA | BT057990.1 | - | 1 - 2 | 1496 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |