Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260351.1 | NP_001247280.1 | 17 - 18 | 12154 | 77 | |
NM_001275974.1 | NP_001262903.1 | 16 - 17 | 10960 | 77 | ||
NM_001275975.1 | NP_001262904.1 | 16 - 17 | 12124 | 77 | ||
NM_001300571.1 | NP_001287500.1 | 15 - 16 | 10921 | 77 | ||
NM_176556.2 | NP_788733.1 | 16 - 17 | 12127 | 77 | ||
NM_176557.2 | NP_788734.1 | 16 - 17 | 12127 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |