Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo95E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032033.2 | NP_001027204.2 | 11 - 12 | 3064 | 79 | |
NM_001032036.2 | NP_001027207.1 | 11 - 12 | 3080 | 79 | ||
NM_206556.2 | NP_996279.1 | 10 - 11 | 3167 | 79 | ||
GenBank mRNA | AF454351.1 | - | 10 - 11 | 3167 | 79 | |
BT011452.1 | - | 11 - 12 | 3075 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |