Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142954.4 | NP_651211.2 | 1 - 2 | 514 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |