Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170279.3 | NP_733158.1 | 2 | 197 | 143 | |
NM_170280.2 | NP_733159.1 | 1 | 261 | 143 | ||
GenBank mRNA | BT001610.1 | - | 1 | 261 | 143 | |
BT003571.1 | - | 2 | 185 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |