Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057325.3 | NP_476673.1 | 2 | 650 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |