Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299883.1 | NP_001286812.1 | 2 | 2291 | 56 | |
NM_138029.3 | NP_611873.1 | 2 | 2028 | 56 | ||
NM_166651.2 | NP_726404.1 | 2 | 2040 | 56 | ||
GenBank mRNA | BT001698.1 | - | 2 | 2056 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |