Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG33970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038982.2 | NP_001034071.1 | 2 | 778 | 94 | |
NM_001300613.1 | NP_001287542.1 | 2 | 778 | 94 | ||
GenBank mRNA | BT003531.1 | - | 2 | 775 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |