Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG46311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347789.1 | NP_001334677.1 | 3 | 1148 | 72 |
Gene Symbol: Tpst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103503.2 | NP_001096973.1 | 3 | 1148 | 72 | |
NM_001272589.1 | NP_001259518.1 | 2 | 500 | 72 | ||
NM_001272590.1 | NP_001259519.1 | 3 | 1148 | 72 | ||
NM_001272591.1 | NP_001259520.1 | 3 | 1148 | 72 | ||
NM_001272592.1 | NP_001259521.1 | 3 | 1342 | 72 | ||
NM_167374.3 | NP_727717.1 | 2 | 500 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |