Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132013.3 | NP_572241.2 | 2 | 575 | 65 | |
NM_167041.3 | NP_727013.2 | 3 | 661 | 65 | ||
NM_167042.3 | NP_727014.2 | 2 | 716 | 65 | ||
GenBank mRNA | BT011040.1 | - | 2446 | 65 | ||
BT099555.1 | - | 2 | 584 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |