Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015157.4 | NP_001015157.2 | 3 - 4 | 362 | 74 | |
NM_001015158.4 | NP_001015158.1 | 3 - 4 | 362 | 74 | ||
NM_001015160.4 | NP_001015160.2 | 3 - 4 | 362 | 74 | ||
NM_001015161.4 | NP_001015161.2 | 3 - 4 | 362 | 74 | ||
NM_001015162.4 | NP_001015162.1 | 3 - 4 | 362 | 74 | ||
NM_001110995.2 | NP_001104465.1 | 3 - 4 | 564 | 74 | ||
NM_001201439.2 | NP_001188368.1 | 4 - 5 | 474 | 74 | ||
NM_001201440.1 | NP_001188369.1 | 3 - 4 | 564 | 74 | ||
GenBank mRNA | BT120108.1 | - | 3 - 4 | 319 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |