Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141189.2 | NP_649446.1 | 2 | 533 | 67 | |
GenBank mRNA | BT120015.1 | - | 1 | 461 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |